Skalpelblade
Nr. 22
Supplier: SWANN MORTON
Jump to:
Product Details & Documents
Kulstofstål.
- Available in sterile individual foil packs or non sterile foil packs of five
- Quick and easy to assemble
Specifications
- Model:Nr. 22
- Seq:09