Orders LinkContactUsLinkComponent
Skalpelblade
Skalpelblade
Nr.10A
Katalog # 233-0101
Leverandør:   SWANN MORTON
Skalpelblade
Katalog # 233-0101
Leverandør:  SWANN MORTON
Skalpelblade
Skalpelblade
Nr.10A
Price
410,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Kulstofstål.
  • Available in sterile individual foil packs or non sterile foil packs of five
  • Quick and easy to assemble
Specifikationer
  • Model:
    Nr.10A
  • Seq:
    04