Methylcyclohexan for synthesis, Sigma-Aldrich®
Katalog # 8.06147.2500
Leverandør: Merck
CAS nummer: 108-87-2
Gå til:
Product Details & Documents
- CAS nummer:108-87-2
- Formel:C₆H₁₁CH₃
- MDL nummer:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:13,06074
- Kogende Pt:101 °C (1013 hPa)
- Smeltende Pt:–126 °C
- Tæthed:0,7693 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Opbevaringstemperatur:Stue temperatur
- FN:2296
- ADR:3,II
- :M-Clarity™ quality level = MQ200
- EINECS:203-624-3
Documents
Certifikater
Søg efter partinummerMERC8.06147.1000_TDS_EN ProductPage_806147
Produktspecifikation
msds_806147
SDS
Specifikationer
Specifikationstestresultater
- Solubility:0.014 g/l (20 °C)
- Melting Point:-126 °C
- Molar Mass:98.19 g/mol
- Boiling Point:100.9 °C (1013 hPa)
- Vapor Pressure:48 hPa (20 °C)
- Flash Point:-4 °C
- Explosion Limit:1.1 - 6.7 %(V)
- Density:0.77 g/cm3 (20 °C)
- Ignition Point:260 °C
- Hill Formula:C7H14
- Chemical Formula:C6H11CH3
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



