Methylcyclohexan for synthesis, Sigma-Aldrich®
Catalog # 8.06147.1000
Supplier: Merck
CAS Number: 108-87-2
Jump to:
Product Details & Documents
- CAS Number:108-87-2
- Formula:C₆H₁₁CH₃
- MDL Number:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:13,06074
- Boiling Pt:101 °C (1013 hPa)
- Melting Pt:–126 °C
- Density:0,7693 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Stue temperatur
- UN:2296
- ADR:3,II
- :M-Clarity™ quality level = MQ200
- EINECS:203-624-3
Documents
Certificates
Search by Lot NumberMERC8.06147.1000_TDS_EN ProductPage_806147
Product specification
msds_806147
SDS
Specifications
Specification Test Results
- Solubility:0.014 g/l (20 °C)
- Melting Point:-126 °C
- Molar Mass:98.19 g/mol
- Boiling Point:100.9 °C (1013 hPa)
- Vapor Pressure:48 hPa (20 °C)
- Flash Point:-4 °C
- Explosion Limit:1.1 - 6.7 %(V)
- Density:0.77 g/cm3 (20 °C)
- Ignition Point:260 °C
- Hill Formula:C7H14
- Chemical Formula:C6H11CH3
Specifications
- Pakning:Glass bottle
- Pakn:1 L



