Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Methylcyclohexan for synthesis, Sigma-Aldrich®

Glass bottle, 100 mL
Katalog # 8.06147.0100
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Merck
Methylcyclohexan for synthesis, Sigma-Aldrich®
Katalog # 8.06147.0100
Leverandør:  Merck
CAS nummer:  108-87-2
Methylcyclohexan for synthesis, Sigma-Aldrich®
Methylcyclohexan for synthesis, Sigma-Aldrich®

Glass bottle, 100 mL

CAS nummer:  108-87-2
Price
305,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
  • CAS nummer:108-87-2
  • Formel:C₆H₁₁CH₃
  • MDL nummer:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:13,06074
  • Kogende Pt:101 °C (1013 hPa)
  • Smeltende Pt:–126 °C
  • Tæthed:0,7693 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:2296
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-624-3
Specifikationer

Specifikationstestresultater

  • Solubility:
    0.014 g/l (20 °C)
  • Melting Point:
    -126 °C
  • Molar Mass:
    98.19 g/mol
  • Boiling Point:
    100.9 °C (1013 hPa)
  • Vapor Pressure:
    48 hPa (20 °C)
  • Flash Point:
    -4 °C
  • Explosion Limit:
    1.1 - 6.7 %(V)
  • Density:
    0.77 g/cm3 (20 °C)
  • Ignition Point:
    260 °C
  • Hill Formula:
    C7H14
  • Chemical Formula:
    C6H11CH3

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    100 mL

Specifikationer