Plader, 1536 brønde, Nunc™
1536-brøndsplade, hvid, hvid, 1536, -
Katalog # 732-2710
Leverandør: Thermo Fisher Scientific
Plader, 1536 brønde, Nunc™
Katalog # 732-2710
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
PS, non treated, without lid. Designed for ultra-high throughput screening applications.
- Standard microplate footprint
- Available non treated in black, white or clear
- Low profile lid available separately
Working range: 1 to 10 µl.
Specifikationer
- Ant. brønde:1536
- Steril:-
- Farve:hvid
- Beskrivelse:1536-brøndsplade, hvid
- Cat. No.:732-2710
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning