Orders LinkContactUsLinkComponent
Plader, 1536 brønde, Nunc™
Plader, 1536 brønde, Nunc™
1536-brøndsplade, sort, sort, 1536, -
Katalog # 732-2709
   Lav fremstillingspåvirkning
Leverandør:   Thermo Fisher Scientific
Plader, 1536 brønde, Nunc™
Katalog # 732-2709
   Lav fremstillingspåvirkning
Plader, 1536 brønde, Nunc™
Plader, 1536 brønde, Nunc™
1536-brøndsplade, sort, sort, 1536, -
Price
57.070,00 kr
Log ind for tilgængelighed
1 * 90 Items
Product Details & Documents
PS, non treated, without lid. Designed for ultra-high throughput screening applications.
  • Standard microplate footprint
  • Available non treated in black, white or clear
  • Low profile lid available separately
Working range: 1 to 10 µl.
Specifikationer
  • Ant. brønde:
    1536
  • Steril:
    -
  • Farve:
    sort
  • Beskrivelse:
    1536-brøndsplade, sort
  • Cat. No.:
    732-2709
  • Miljøcertificering:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Lav fremstillingspåvirkning