Buffer solutions in sachets, Certipur®, Supelco®
Certipur® buffer solution, set 2 (pH 4,01, 7,00 and 10,00), 4,01/7,00/10,00, Sachet, 30×30 ml, 30
Leverandør: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Katalog # 1.99006.0001
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
Til kalibrering af pH-metre.
- Practical 30 ml sachets - one use per sachet - no risk of contamination
- Solutions are always fresh and ready to use
- Traceable to SRM from NIST and PTB
- Reference temperature: +25 °C
- Precision: Deviation is max. ±0,015 pH units
pH value is measured with a combined glass electrode after 5-point calibration according to DIN 19268 with reference buffer solutions according to DIN 19266, IUPAC, NIST, Ph. Eur. and USP.
Specifikationer
Specifikationer
- Pakning:Sachet
- Pakn:30
- Beskrivelse:Certipur® buffer solution, set 2 (pH 4,01, 7,00 and 10,00)
- Pk Info:30×30 ml
- pH-værdi:4,01/7,00/10,00
- Cat. No.:1.99006.0001
- M-Clarity quality level:MQ100



