Buffer solutions in sachets, Certipur®, Supelco®
CertiPUR® buffer solution, pH 4,01, potassium hydrogen phthalate, 4,01, Phthalat, Sachet, 30×30 ml, 30
Leverandør: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Katalog # 1.99001.0001
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
Til kalibrering af pH-metre.
- Practical 30 ml sachets - one use per sachet - no risk of contamination
- Solutions are always fresh and ready to use
- Traceable to SRM from NIST and PTB
- Reference temperature: +25 °C
- Precision: Deviation is max. ±0,015 pH units
pH value is measured with a combined glass electrode after 5-point calibration according to DIN 19268 with reference buffer solutions according to DIN 19266, IUPAC, NIST, Ph. Eur. and USP.
Documents
Certifikater
Søg efter partinummerCertipur Merck Buffer Calibration Standards for pH Measurements Merck Pharma Segment Brochure
Product Literature
MERC1.99001.0001_TDS_EN MERC1.99002.0001_TDS_EN MERC1.99003.0001_TDS_EN MERC1.99004.0001_TDS_EN MERC1.99005.0001_TDS_EN
Datasheet
More
Spec MERC1.99002.0001.pdf
Product specification
Specifikationer
Specifikationer
- Pakning:Sachet
- pH buffer type:Phthalat
- Pakn:30
- Beskrivelse:CertiPUR® buffer solution, pH 4,01, potassium hydrogen phthalate
- Pk Info:30×30 ml
- pH-værdi:4,01
- Cat. No.:1.99001.0001
- M-Clarity quality level:MQ100
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



