Ion chromatography Multi-Element standards, Supelco®
Ion chromatography IC anion multi-element standard solution I, (5 elements in water) 100 mg/l: F , 250 mg/l: Cl, 500 mg/l: NO3 / 500 mg/l: SO4 / 1000 mg/l: PO4 Certipur®, 500 mL
Leverandør: Merck
Ion chromatography Multi-Element standards, Supelco®
Katalog # 1.70398.0500
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
All ion chromatography standards are traceable to NIST standard reference materials.
Specifikationer
Specifikationer
- Pakn:500 mL
- Beskrivelse:Ion chromatography IC anion multi-element standard solution I, (5 elements in water) 100 mg/l: F , 250 mg/l: Cl, 500 mg/l: NO<sub>3</sub> / 500 mg/l: SO<sub>4</sub> / 1000 mg/l: PO<sub>4</sub> Certipur®
- Cat. No.:1.70398.0500
- M-Clarity quality level:MQ100