Orders LinkContactUsLinkComponent
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography IC anion multi-element standard solution I, (5 elements in water) 100 mg/l: F , 250 mg/l: Cl, 500 mg/l: NO3 / 500 mg/l: SO4 / 1000 mg/l: PO4 Certipur®, 500 mL
Katalog # 1.70398.0500
Leverandør:   Merck
Ion chromatography Multi-Element standards, Supelco®
Katalog # 1.70398.0500
Leverandør:  Merck
CAS nummer:  
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography IC anion multi-element standard solution I, (5 elements in water) 100 mg/l: F , 250 mg/l: Cl, 500 mg/l: NO3 / 500 mg/l: SO4 / 1000 mg/l: PO4 Certipur®, 500 mL
CAS nummer:  
Price
1.360,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
All ion chromatography standards are traceable to NIST standard reference materials.
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Beskrivelse:
    Ion chromatography IC anion multi-element standard solution I, (5 elements in water) 100 mg/l: F , 250 mg/l: Cl, 500 mg/l: NO<sub>3</sub> / 500 mg/l: SO<sub>4</sub> / 1000 mg/l: PO<sub>4</sub> Certipur®
  • Cat. No.:
    1.70398.0500
  • M-Clarity quality level:
    MQ100
Specifikationer