Perfect DNA™ markører
Molecular size markers for routine use in plasmid cloning, the Perfect DNA™ markers contain sets of DNA species having convenient, easy-to-remember sizes for agarose gel analysis. The markers have uniform band intensities except for the indicated reference bands, which are useful for instant band identification.
- Easy-to-remember sizes
- Uniform band intensities
- Instant band identification
Product components:
1 ml Perfect DNA Markers i 1X DNA Gel Loading buffer, til markørstørrelse 0,05 10 kbp (Art.nr 70540-3) eller 500 μl Perfect DNA Markers i 1X DNA Gel Loading buffer, til markørstørrelse 0,1 12 og 0,5 12 kbp (Art.nr 70087-3 og 69002-3).
1 ml 6X DNA Gel Loading buffer.
Delivery information: Hvert rør med markører indeholder nok til 100 baner. Et ekstra rør med 6X DNA Gel Loading buffer er inkluderet.
- Pakn:1
- Beskrivelse:Perfect DNA™ markører, 0,1-12 kbp