Orders LinkContactUsLinkComponent
Perfect DNA™ markører
Perfect DNA™ markører
Perfect DNA™ markører, 0,5-12 kbp, 1
Katalog # 69002-3
Perfect DNA™ markører
Katalog # 69002-3
CAS nummer:  
Perfect DNA™ markører
Perfect DNA™ markører, 0,5-12 kbp, 1
CAS nummer:  
Price
1.290,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Molecular size markers for routine use in plasmid cloning, the Perfect DNA™ markers contain sets of DNA species having convenient, easy-to-remember sizes for agarose gel analysis. The markers have uniform band intensities except for the indicated reference bands, which are useful for instant band identification.
  • Easy-to-remember sizes
  • Uniform band intensities
  • Instant band identification
Product components:
1 ml Perfect DNA Markers i 1X DNA Gel Loading buffer, til markørstørrelse 0,05 10 kbp (Art.nr 70540-3) eller 500 μl Perfect DNA Markers i 1X DNA Gel Loading buffer, til markørstørrelse 0,1 12 og 0,5 12 kbp (Art.nr 70087-3 og 69002-3).
1 ml 6X DNA Gel Loading buffer.
Leveringsinformation: Hvert rør med markører indeholder nok til 100 baner. Et ekstra rør med 6X DNA Gel Loading buffer er inkluderet.
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Perfect DNA™ markører, 0,5-12 kbp