Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sorbents, LiChrospher®

Sorbents, LiChrospher®

LiChrospher® 60 RP-selected B, 5 µm, 01, 10 g
Catalog # 1.19641.0010
Supplier:   Merck
Sorbents, LiChrospher®
Catalog # 1.19641.0010
Supplier:  Merck
CAS Number:  
Sorbents, LiChrospher®
Sorbents, LiChrospher®

LiChrospher® 60 RP-selected B, 5 µm, 01, 10 g

CAS Number:  
Price
18.540,00 kr
Log in for availability
1 * 10 g
Product Details & Documents
Specifications

Specifications

  • Partikelstørrelse:
    5 µm
  • Pakn:
    10 g
  • Porestørrelse:
    01
  • Beskrivelse:
    LiChrospher® 60 RP-selected B

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.