Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Sorbents, LiChrospher®

LiChrospher® Si 60, 5 µm, 13, 10 g
Katalog # 1.19640.0010
Leverandør:   Merck
Sorbents, LiChrospher®
Katalog # 1.19640.0010
Leverandør:  Merck
CAS nummer:  
Sorbents, LiChrospher®
Sorbents, LiChrospher®

LiChrospher® Si 60, 5 µm, 13, 10 g

CAS nummer:  
Price
5.440,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
Specifikationer

Specifikationer

  • Partikelstørrelse:
    5 µm
  • Pakn:
    10 g
  • Porestørrelse:
    13
  • Beskrivelse:
    LiChrospher® Si 60

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.