Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sorbents, LiChrospher®

Sorbents, LiChrospher®

LiChrospher® 100 CN, 5 µm, 03, 10 g
Catalog # 1.19638.0010
Supplier:   Merck
Sorbents, LiChrospher®
Catalog # 1.19638.0010
Supplier:  Merck
CAS Number:  
Sorbents, LiChrospher®
Sorbents, LiChrospher®

LiChrospher® 100 CN, 5 µm, 03, 10 g

CAS Number:  
Price
14.500,00 kr
Log in for availability
1 * 10 g
Product Details & Documents
Specifications

Specifications

  • Partikelstørrelse:
    5 µm
  • Pakn:
    10 g
  • Porestørrelse:
    03
  • Beskrivelse:
    LiChrospher® 100 CN

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.