Molecular models of elements, Molymod®
Phosphorus, lilla, 5, 23 mm
Katalog # SPIRMA-510
Marketsource
Leverandør: SPIRING ENTERPRISES
Molecular models of elements, Molymod®
Katalog # SPIRMA-510
Leverandør: SPIRING ENTERPRISES
Marketsource
Gå til:
Product Details & Documents
Molymod is the original, unique, dual-scale system of high quality molecular models.
Specifikationer
- Farve:lilla
- Beskrivelse:Phosphorus
- Antal huller:5
- Ø:23 mm