Reflectoquant® teststrimler, Supelco®
Reflectoquant® 10 plus phosphattest, 0,1 - 5,0 mg/l PO₄, Phosphormolybdenum blue, RQflex® 10 Plus, 100 Tests
Leverandør: Merck
Product Details & Documents
Test strips for the reflectometers RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Documents
Specifikationer
Specifikationer
- Kemisk metode:Phosphormolybdenum blue
- Pakn:100 Tests
- Beskrivelse:Reflectoquant® 10 plus phosphattest
- Til enhed:RQflex® 10 Plus
- Område:0,1 - 5,0 mg/l PO₄
- Cat. No.:1.17942.0001
- M-Clarity quality level:MQ100
Specifikationer
Complementary Products
Produktinformation
- Katalog #BeskrivelseModelTilgængelighedPrice
- 1.17246.0001Reflectometer for test strips onlyRQflex 20Log ind for tilgængelighed1 * 1 Items - 10.580,00 kr