Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Reflectoquant® teststrimler, Supelco®

Reflectoquant® teststrimler, Supelco®

pH-test, pH 4,0 - 9,0, Mixed indicator, 50 Tests
Katalog # 1.16996.0001
Leverandør:   Merck
Reflectoquant® teststrimler, Supelco®
Katalog # 1.16996.0001
Leverandør:  Merck
CAS nummer:  
Reflectoquant® teststrimler, Supelco®
Reflectoquant® teststrimler, Supelco®

pH-test, pH 4,0 - 9,0, Mixed indicator, 50 Tests

CAS nummer:  
Price
476,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.

  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
Specifikationer

Specifikationer

  • Kemisk metode:
    Mixed indicator
  • Pakn:
    50 Tests
  • Beskrivelse:
    pH-test
  • Område:
    pH 4,0 - 9,0
  • Cat. No.:
    1.16996.0001
  • M-Clarity quality level:
    MQ100

Specifikationer

Complementary Products

Produktinformation

  • Katalog #
    Beskrivelse
    Model
    Tilgængelighed
    Price
  • 1.17246.0001
    Reflectometer for test strips only
    RQflex 20
    Log ind for tilgængelighed
    1 * 1 Items - 10.580,00 kr