Reflectoquant® teststrimler, Supelco®
Fosfat test, 5 - 120 mg/l PO₄, Phosphormolybdenum blue, RQflex® 20, 50 Tests
: Merck
Test strips for the reflectometers RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
- Kemisk metode:Phosphormolybdenum blue
- Pakn:50 Tests
- Beskrivelse:Fosfat test
- Til enhed:RQflex® 20
- Område:5 - 120 mg/l PO₄
- Cat. No.:1.16978.0001
- M-Clarity quality level:MQ100