Orders LinkContactUsLinkComponent
Reflectoquant® teststrimler, Supelco®
Ammoniumtest, 20 - 180 mg/l NH₄, Nesslers reagent, 50 Tests
  1.16977.0001
 :   Merck
Reflectoquant® teststrimler, Supelco®
  1.16977.0001
 :  Merck
 :  
Reflectoquant® teststrimler, Supelco®
Reflectoquant® teststrimler, Supelco®
Ammoniumtest, 20 - 180 mg/l NH₄, Nesslers reagent, 50 Tests
 :  
964,00 kr
1 * 1 Each
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
  • Kemisk metode:
    Nesslers reagent
  • Pakn:
    50 Tests
  • Beskrivelse:
    Ammoniumtest
  • Område:
    20 - 180 mg/l NH₄
  • Cat. No.:
    1.16977.0001
  • M-Clarity quality level:
    MQ100
Complementary Products
  •  
    Beskrivelse
    Model
  • 1.17246.0001
    Reflectometer for test strips only
    RQflex 20
    1 * 1 Items - 10.580,00 kr