Reflectoquant® teststrimler, Supelco®
Ammoniumtest, 20 - 180 mg/l NH₄, Nesslers reagent, 50 Tests
: Merck
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
- Kemisk metode:Nesslers reagent
- Pakn:50 Tests
- Beskrivelse:Ammoniumtest
- Område:20 - 180 mg/l NH₄
- Cat. No.:1.16977.0001
- M-Clarity quality level:MQ100