Orders LinkContactUsLinkComponent
Testkits, calciumtest, til Spectroquant®, Supelco®
Testkits, calciumtest, til Spectroquant®, Supelco®
Calciumtest, 5 - 160 mg/l Ca, 7 - 224 CaO, 12 - 400 mg/l CaCO₃, 1,0 mg/l - 15,0 mg/l Ca, 1,4 - 21,0 mg/l CaO, 2,5 - 37,5 CaCO₃, Glyoxal-bis(2-hydroxyaniline), prove, Nova 60, Multy, Move, 100 Tests
Catalog # 1.14815.0001
Supplier:   Merck
Testkits, calciumtest, til Spectroquant®, Supelco®
Catalog # 1.14815.0001
Supplier:  Merck
CAS Number:  
Testkits, calciumtest, til Spectroquant®, Supelco®
Testkits, calciumtest, til Spectroquant®, Supelco®
Calciumtest, 5 - 160 mg/l Ca, 7 - 224 CaO, 12 - 400 mg/l CaCO₃, 1,0 mg/l - 15,0 mg/l Ca, 1,4 - 21,0 mg/l CaO, 2,5 - 37,5 CaCO₃, Glyoxal-bis(2-hydroxyaniline), prove, Nova 60, Multy, Move, 100 Tests
CAS Number:  
Price
1.980,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits.
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Specifikationer
Specifikationer
  • Kemisk metode:
    Glyoxal-bis(2-hydroxyaniline)
  • Pakn:
    100 Tests
  • Beskrivelse:
    Calciumtest
  • Til enhed:
    prove, Nova 60, Multy, Move
  • Område:
    5 - 160 mg/l Ca, 7 - 224 CaO, 12 - 400 mg/l CaCO₃, 1,0 mg/l - 15,0 mg/l Ca, 1,4 - 21,0 mg/l CaO, 2,5 - 37,5 CaCO₃
  • Cat. No.:
    1.14815.0001
  • M-Clarity quality level:
    MQ100