Testkits, calciumtest, til Spectroquant®, Supelco®
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits.
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Documents
Specifikationer
Specifikationer
- Kemisk metode:Glyoxal-bis(2-hydroxyaniline)
- Pakn:100 Tests
- Beskrivelse:Calciumtest
- Til enhed:prove, Nova 60, Multy, Move
- Område:5 - 160 mg/l Ca, 7 - 224 CaO, 12 - 400 mg/l CaCO₃, 1,0 mg/l - 15,0 mg/l Ca, 1,4 - 21,0 mg/l CaO, 2,5 - 37,5 CaCO₃
- Cat. No.:1.14815.0001
- M-Clarity quality level:MQ100