Test kits, potassium cell test, Spectroquant®, Supelco®
Kalium celle test, 5,0 - 50,0 mg/L K, Kalignost, Prove, Nova 30/60, Multy, Move, 25 Tests
Leverandør: Merck
Test kits, potassium cell test, Spectroquant®, Supelco®
Katalog # 1.14562.0001
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Documents
Specifikationer
Specifikationer
- Kemisk metode:Kalignost
- Pakn:25 Tests
- Beskrivelse:Kalium celle test
- Til enhed:Prove, Nova 30/60, Multy, Move
- Område:5,0 - 50,0 mg/L K
- Cat. No.:1.14562.0001
- M-Clarity quality level:MQ100