Supelco® Spectroquant® Test Kits, Phosphate
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 Denne metode er officielt anerkendt af USEPA som en alternativ metode til undersøgelse af følgende:
*7-1 spildevand
*7-2 drikkevand
*7-3 hhv. drikke- og spildevand
Documents
Certifikater
Søg efter partinummerMERC1.00474.0001_TDS_EN
Datablad
Specifikationer
Specifikationer
- Kemisk metode:Vanadato-molybdate
- Pakn:25 Tests
- Beskrivelse:Phosphatkuvettetest
- Til enhed:Prove, Nova 30/60, Move
- Område:0,5 - 25,0 mg/L PO₄ - P, 1,5 - 76,7 mg/L PO₄, 1,1 - 57,3 mg/L P₂O₅
- Cat. No.:1.14546.0001
- M-Clarity quality level:MQ100