Orders LinkContactUsLinkComponent
Supelco® Spectroquant® Test Kits, Phosphate
Supelco® Spectroquant® Test Kits, Phosphate
Phosphate cell test, 0,05 - 5,00 mg/L PO₄ - P, 0,2 - 15,3 PO₄, 0,11 - 11,46 P₂O₅, Phosphormolybdenum blue, Prove, Nova 30/60, Multy, Move, 25 Tests
Katalog # 1.14543.0001
Leverandør:   Merck
Supelco® Spectroquant® Test Kits, Phosphate
Katalog # 1.14543.0001
Leverandør:  Merck
CAS nummer:  
Supelco® Spectroquant® Test Kits, Phosphate
Supelco® Spectroquant® Test Kits, Phosphate
Phosphate cell test, 0,05 - 5,00 mg/L PO₄ - P, 0,2 - 15,3 PO₄, 0,11 - 11,46 P₂O₅, Phosphormolybdenum blue, Prove, Nova 30/60, Multy, Move, 25 Tests
CAS nummer:  
Price
1.760,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 Denne metode er officielt anerkendt af USEPA som en alternativ metode til undersøgelse af følgende:
*7-1 spildevand
*7-2 drikkevand
*7-3 hhv. drikke- og spildevand
Specifikationer
Specifikationer
  • Kemisk metode:
    Phosphormolybdenum blue
  • Pakn:
    25 Tests
  • Beskrivelse:
    Phosphate cell test
  • Til enhed:
    Prove, Nova 30/60, Multy, Move
  • Område:
    0,05 - 5,00 mg/L PO₄ - P, 0,2 - 15,3 PO₄, 0,11 - 11,46 P₂O₅
  • Cat. No.:
    1.14543.0001
  • M-Clarity quality level:
    MQ100
Specifikationer