Ion chromatography Multi-Element standards, Supelco®
Ion chromatography anion multi-element standard solution I, (3 elements in water) 1000 mg/l: F, Br, PO4 Certipur®, 500 mL
: Merck
All ion chromatography standards are traceable to NIST standard reference materials.
Documents
- Pakn:500 mL
- Beskrivelse:Ion chromatography anion multi-element standard solution I, (3 elements in water) 1000 mg/l: F, Br, PO<sub>4 </sub>Certipur®
- Cat. No.:1.11437.0500
- M-Clarity quality level:MQ100