Pipette stand, 337
7 enkeltkanalpipetter, Polyamid, lysegrå
Katalog # 613-4831
Leverandør: Socorex Isba
Gå til:
Product Details & Documents
This space-saving pipette stand made from PA holds up to seven single channel pipettes of most brands.
- Made of sturdy polyamid material, easy to clean
- Very stable with anti-slip pads
- Available in five pastel colours
Specifikationer
- Til:7 enkeltkanalpipetter
- Farve:lysegrå
- Materiale:Polyamid