Orders LinkContactUsLinkComponent
Acrylamid, Millipore®
Acrylamid, Millipore®
Glass bottle, 100 g
Catalog # 1.10784.0100
Toxic
Health
Supplier:   Merck
Acrylamid, Millipore®
Catalog # 1.10784.0100
Supplier:  Merck
CAS Number:  79-06-1
Acrylamid, Millipore®
Acrylamid, Millipore®
Glass bottle, 100 g
CAS Number:  79-06-1
Price
393,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:232 °C (1013 hPa)
  • Melting Pt:84,5 °C
  • Density:1,122 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Køleskab
  • EINECS:201-173-7
  • ADR:6.1,
  •  :M-Clarity™ quality level = MQ200
  • UN:2074
Documents
Specifications
Specifications
  • Pakning:
    Glass bottle
  • Pakn:
    100 g