Ion chromatography Multi-Element standards, Supelco®
Ion chromatography cation multi-element standard solution I, (9 elements in diluted nitic acid) 100 mg/l: Li, Na, K, NH4, Mn, Ca, Mg, Sr, Ba Certipur®, 100 mL
Leverandør: Merck
Ion chromatography Multi-Element standards, Supelco®
Katalog # 1.10322.0100
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
All ion chromatography standards are traceable to NIST standard reference materials.
Specifikationer
Specifikationer
- Pakn:100 mL
- Beskrivelse:Ion chromatography cation multi-element standard solution I, (9 elements in diluted nitic acid) 100 mg/l: Li, Na, K, NH<sub>4</sub>, Mn, Ca, Mg, Sr, Ba<sub> </sub>Certipur®
- Cat. No.:1.10322.0100
- M-Clarity quality level:MQ100