Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Preparative sorbents, LiChroprep®

Preparative sorbents, LiChroprep®

LiChroprep® Si 60, 15 - 25 µm, 13, 25 kg
Katalog # 1.09336.9025
Preparative sorbents, LiChroprep®
Katalog # 1.09336.9025
CAS nummer:  
Preparative sorbents, LiChroprep®

LiChroprep® Si 60, 15 - 25 µm, 13, 25 kg

CAS nummer:  
Price
Spørg for pris
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents

LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.

  • Reproducible homogeneous silica gel matrix
  • Narrow, well defined particle size distribution for high performance and high permeability
  • Excellent selectivity and efficiency
  • Large number of applications
  • Large batch size
  • Comprehensive regulatory documents are available

LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.

Specifikationer

Specifikationer

  • Partikelstørrelse:
    15 - 25 µm
  • Pakn:
    25 kg
  • Porestørrelse:
    13
  • Beskrivelse:
    LiChroprep® Si 60

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.