Preparative sorbents, LiChroprep®
LiChroprep® RP-18, 25 - 40 µm, 05, 500 g
Supplier: MERCK PRODUCTION CHEMICALS
Preparative sorbents, LiChroprep®
Catalog # 1.09303.0500
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Jump to:
Product Details & Documents
LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.
- Reproducible homogeneous silica gel matrix
- Narrow, well defined particle size distribution for high performance and high permeability
- Excellent selectivity and efficiency
- Large number of applications
- Large batch size
- Comprehensive regulatory documents are available
LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.
Documents
Specifications
Specifications
- Partikelstørrelse:25 - 40 µm
- Pakn:500 g
- Porestørrelse:05
- Beskrivelse:LiChroprep® RP-18



