Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L(+)-Valin, Millipore®

L(+)-Valin, Millipore®

Plastic drum, 10 kg
Katalog # 1.08495.9010
Leverandør:   Merck
L(+)-Valin, Millipore®
Katalog # 1.08495.9010
Leverandør:  Merck
CAS nummer:  72-18-4
L(+)-Valin, Millipore®

Plastic drum, 10 kg

CAS nummer:  72-18-4
Price
Spørg for pris
Log ind for tilgængelighed
1 * 10 kg
Product Details & Documents
Synonymer:  H-Val-OH, Valine, (S)-(+)-Valine
  • CAS nummer:72-18-4
  • Formel:C₅H₁₁NO₂
  • MDL nummer:MFCD00064220
  • Molecular Weight:117.15 g/mol
  • Merck Index:13,09458
  • Kogende Pt:217 °C (1013 hPa)
  • Smeltende Pt:86 °C
  • Tæthed:1,284 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-773-6

Documents

Specifikationer

Specifikationstestresultater

  • Ammonium (NH4):
    0.01% max.
  • Foreign Amino Acids:
    0.3% max.
  • Heavy Metals (as Pb):
    0.001% max.
  • Melting Point:
    315°C

Specifikationer

  • Pakning:
    Plastic drum
  • Pakn:
    10 kg

Specifikationer