L(+)-Valin, Millipore®
Plastic bottle, 100 g
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: H-Val-OH, Valine, (S)-(+)-Valine
- CAS nummer:72-18-4
- Formel:C₅H₁₁NO₂
- MDL nummer:MFCD00064220
- Molecular Weight:117.15 g/mol
- Merck Index:13,09458
- Kogende Pt:217 °C (1013 hPa)
- Smeltende Pt:86 °C
- Tæthed:1,284 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-773-6
Documents
Specifikationer
Specifikationstestresultater
- Ammonium (NH4):0.01% max.
- Foreign Amino Acids:0.3% max.
- Heavy Metals (as Pb):0.001% max.
- Melting Point:315°C
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g



