Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L(+)-Valin, Millipore®

L(+)-Valin, Millipore®

Plastic bottle, 25 g
Katalog # 1.08495.0025
Leverandør:   Merck
L(+)-Valin, Millipore®
Katalog # 1.08495.0025
Leverandør:  Merck
CAS nummer:  72-18-4
L(+)-Valin, Millipore®

Plastic bottle, 25 g

CAS nummer:  72-18-4
Price
405,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  H-Val-OH, Valine, (S)-(+)-Valine
  • CAS nummer:72-18-4
  • Formel:C₅H₁₁NO₂
  • MDL nummer:MFCD00064220
  • Molecular Weight:117.15 g/mol
  • Merck Index:13,09458
  • Kogende Pt:217 °C (1013 hPa)
  • Smeltende Pt:86 °C
  • Tæthed:1,284 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-773-6

Documents

Certifikater
Søg efter partinummer
Specifikationer

Specifikationstestresultater

  • Ammonium (NH4):
    0.01% max.
  • Foreign Amino Acids:
    0.3% max.
  • Heavy Metals (as Pb):
    0.001% max.
  • Melting Point:
    315°C

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.