Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L(-)-Tryptophan, Millipore®

L(-)-Tryptophan, Millipore®

Plastic bottle, 100 g
Katalog # 1.08374.0100
Leverandør:   Merck
L(-)-Tryptophan, Millipore®
Katalog # 1.08374.0100
Leverandør:  Merck
CAS nummer:  73-22-3
L(-)-Tryptophan, Millipore®
L(-)-Tryptophan, Millipore®

Plastic bottle, 100 g

CAS nummer:  73-22-3
Price
1.500,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
  • CAS nummer:73-22-3
  • Formel:C₁₁H₁₂N₂O₂
  • MDL nummer:MFCD00064340
  • Molecular Weight:204.23 g/mol
  • Merck Index:12,09929
  • Smeltende Pt:290 °C
  • Tæthed:1,34 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-795-6
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.