L(-)-Tryptophan, Millipore®
Plastic bottle, 10 g
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
- CAS nummer:73-22-3
- Formel:C₁₁H₁₂N₂O₂
- MDL nummer:MFCD00064340
- Molecular Weight:204.23 g/mol
- Merck Index:12,09929
- Smeltende Pt:290 °C
- Tæthed:1,34 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-795-6
Documents
Certifikater
Søg efter partinummerMERC1.08374.0010_TDS_EN ProductPage_108374 ProductPage_108374
Produktspecifikation
Specifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:10 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



