Phenol rød ACS
Glass bottle, 100 g
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS nummer:143-74-8
- Formel:C₁₉H₁₄O₅S
- MDL nummer:MFCD00003552
- Molecular Weight:354.38 g/mol
- Merck Index:13,07329
- Smeltende Pt:> 300 °C
- :M-Clarity™ quality level = MQ200
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-609-7
Specifikationer
Specifikationstestresultater
- Solubility:0.77 g/l (100 °C)
- Melting Point:300 °C
- Molar Mass:354.38 g/mol
- Bulk Density:200 - 300 kg/m3
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



