Phenol rød ACS
Glass bottle, 25 g
: Merck
: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- :143-74-8
- :C₁₉H₁₄O₅S
- :MFCD00003552
- :354.38 g/mol
- :13,07329
- :> 300 °C
- :M-Clarity™ quality level = MQ200
- :Stue temperatur
- :205-609-7
Documents
- Solubility:0.77 g/l (100 °C)
- Melting Point:300 °C
- Molar Mass:354.38 g/mol
- Bulk Density:200 - 300 kg/m3
- Pakning:Glass bottle
- Pakn:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



