Natriumpyruvat, Millipore®
Plastic bottle, 1 kg
: Merck
: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- :113-24-6
- :CH₃COCOONa
- :MFCD00002586
- :110.04 g/mol
- :220…230 °C
- :1,718 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- :Stue temperatur
- :204-024-4
- Pakning:Plastic bottle
- Pakn:1 kg