Natriumpyruvat, Millipore®
Plastic bottle, 50 g
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS nummer:113-24-6
- Formel:CH₃COCOONa
- MDL nummer:MFCD00002586
- Molecular Weight:110.04 g/mol
- Smeltende Pt:220…230 °C
- Tæthed:1,718 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-024-4
Documents
Specifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:50 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



