Orders LinkContactUsLinkComponent
Syringes with valves
Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 100 ml, 100MR-VLLMA-GT
Catalog # SGEA009770
Syringes with valves
Catalog # SGEA009770
Syringes with valves
Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 100 ml, 100MR-VLLMA-GT
Price
4.340,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Specifications
  • Model:
    100MR-VLLMA-GT
  • Volumen:
    100 ml
  • Beskrivelse:
    Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
  • Seq:
    12