Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 100 ml, 100MR-VLLMA-GT
Catalog # SGEA009770
Supplier: Trajan Scientific and Medical
Jump to:
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Specifications
- Model:100MR-VLLMA-GT
- Volumen:100 ml
- Beskrivelse:Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
- Seq:12