Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 10 ml, 10MDR-VLLMA-GT
Katalog # SGEA008970
Leverandør: Trajan Scientific and Medical
Gå til:
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Specifikationer
- Model:10MDR-VLLMA-GT
- Volumen:10 ml
- Beskrivelse:Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
- Seq:09