Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 1 ml, 1MR-VLL-GT
Katalog # SGEA008160
Leverandør: Trajan Scientific and Medical
Gå til:
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Specifikationer
- Model:1MR-VLL-GT
- Volumen:1 ml
- Beskrivelse:Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
- Seq:06