Orders LinkContactUsLinkComponent
Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 1 ml, 1MR-VLL-GT
Katalog # SGEA008160
Syringes with valves
Katalog # SGEA008160
Syringes with valves
Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 1 ml, 1MR-VLL-GT
Price
1.540,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Specifikationer
  • Model:
    1MR-VLL-GT
  • Volumen:
    1 ml
  • Beskrivelse:
    Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
  • Seq:
    06