Orders LinkContactUsLinkComponent
Syringes with valves
Syringe pre-fitted with removable needle and valve, PTFE tipped plunger, 1 ml, 1MR-V-GT
  SGEA008110
Syringes with valves
  SGEA008110
Syringes with valves
Syringes with valves
Syringe pre-fitted with removable needle and valve, PTFE tipped plunger, 1 ml, 1MR-V-GT
1.590,00 kr
1 * 1 Items
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
  • Model:
    1MR-V-GT
  • Volumen:
    1 ml
  • Beskrivelse:
    Syringe pre-fitted with removable needle and valve, PTFE tipped plunger
  • Seq:
    02