Syringes with valves
Syringe pre-fitted with removable needle and valve, PTFE tipped plunger, 1 ml, 1MR-V-GT
SGEA008110
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
- Model:1MR-V-GT
- Volumen:1 ml
- Beskrivelse:Syringe pre-fitted with removable needle and valve, PTFE tipped plunger
- Seq:02