Orders LinkContactUsLinkComponent
Syringes with valves
Syringe pre-fitted with removable needle and valve, PTFE tipped plunger, 0,5 ml, 500R-V-GT
  SGEA007279
Syringes with valves
  SGEA007279
Syringes with valves
Syringes with valves
Syringe pre-fitted with removable needle and valve, PTFE tipped plunger, 0,5 ml, 500R-V-GT
1.450,00 kr
1 * 1 Items
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
  • Model:
    500R-V-GT
  • Volumen:
    0,5 ml
  • Beskrivelse:
    Syringe pre-fitted with removable needle and valve, PTFE tipped plunger
  • Seq:
    01