Syringes with valves
Syringe pre-fitted with removable needle and valve, PTFE tipped plunger, 0,5 ml, 500R-V-GT
SGEA007279
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
- Model:500R-V-GT
- Volumen:0,5 ml
- Beskrivelse:Syringe pre-fitted with removable needle and valve, PTFE tipped plunger
- Seq:01