Natriumhydroxid, pellets, pure, Supelco®
Plastic bottle, 1 kg
Leverandør: Merck
Natriumhydroxid, pellets, pure, Supelco®
Katalog # 1.06462.1000
Leverandør: Merck
CAS nummer: 1310-73-2
Gå til:
Product Details & Documents
Synonymer: Caustic soda
- CAS nummer:1310-73-2
- Formel:NaOH
- MDL nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Kogende Pt:1390 °C (1013 hPa)
- Smeltende Pt:323 °C
- Tæthed:2,13 g/cm³ (20 °C)
- EINECS:215-185-5
- ADR:8,II
- :M-Clarity™ quality level = MQ200
- Opbevaringstemperatur:Stue temperatur
- FN:1823
- NÅ:01-2119457892-27
Documents
Certifikater
Søg efter partinummerMERC1.06462.1000_TDS_EN MERC1.06462.5000_TDS_EN ProductPage_106462
Product specification
msds_106462
SDS
Specifikationer
Specifikationstestresultater
- Solubility:1090 g/l (20 °C)
- Melting Point:323 °C
- Molar Mass:40.00 g/mol
- Boiling Point:1390 °C (1013 hPa)
- Vapor Pressure:(20 °C)
- Density:2.13 g/cm3 (20 °C)
- pH:14 (50 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



