Syringes for CTC analytics systems
Removable needle, 26, 25 µl, 50 mm, 0,47 mm, Side hole, 25R-C/T-GT-5/0.47H *
Katalog # 548-1620
Leverandør: Trajan Scientific and Medical
Gå til:
Product Details & Documents
* gastæt
Specifikationer
- Udv.Ø:0,47 mm
- Måler:26
- Model:25R-C/T-GT-5/0.47H *
- Spidstype:Side hole
- Volumen:25 µl
- Beskrivelse:Removable needle
- Længde:50 mm
- Seq:40