Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES monohydrat, Millipore®

MES monohydrat, Millipore®

Glass bottle, 250 g
Catalog # 1.06126.0250
Supplier:   Merck
MES monohydrat, Millipore®
Catalog # 1.06126.0250
Supplier:  Merck
CAS Number:  145224-94-8
MES monohydrat, Millipore®

Glass bottle, 250 g

CAS Number:  145224-94-8
Price
1.410,00 kr
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  • CAS Number:145224-94-8
  • Formula:C₆H₁₃NO₄S·1H₂O
  • MDL Number:MFCD00149409
  • Molecular Weight:213.25 g/mol
  • Merck Index:13,05929
  • Melting Pt:> 300 °C
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Stue temperatur
  • EINECS:224-632-3
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    250 g

Specifications