Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES monohydrat, Millipore®

MES monohydrat, Millipore®

Glass bottle, 25 g
Katalog # 1.06126.0025
Leverandør:   Merck
MES monohydrat, Millipore®
Katalog # 1.06126.0025
Leverandør:  Merck
CAS nummer:  145224-94-8
MES monohydrat, Millipore®

Glass bottle, 25 g

CAS nummer:  145224-94-8
Price
386,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  • CAS nummer:145224-94-8
  • Formel:C₆H₁₃NO₄S·1H₂O
  • MDL nummer:MFCD00149409
  • Molecular Weight:213.25 g/mol
  • Merck Index:13,05929
  • Smeltende Pt:> 300 °C
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:224-632-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.