Methyl rød natrium salt
Glass bottle, 100 g
: Merck
: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
- :845-10-3
- :C₁₅H₁₄N₃NaO₂
- :MFCD00002426
- :291.28 g/mol
- :13,06142
- :> 180 °C
- :M-Clarity™ quality level = MQ200
- :Stue temperatur
- :212-682-9
- Solubility:800 g/l (25 °C)
- Molar Mass:291.29 g/mol
- Bulk Density:360 kg/m3
- pH:8.2 (10 g/l, H2O, 25 °C)
- Pakning:Glass bottle
- Pakn:100 g



