Methyl rød natrium salt
Glass bottle, 25 g
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
- CAS nummer:845-10-3
- Formel:C₁₅H₁₄N₃NaO₂
- MDL nummer:MFCD00002426
- Molecular Weight:291.28 g/mol
- Merck Index:13,06142
- Smeltende Pt:> 180 °C
- :M-Clarity™ quality level = MQ200
- Opbevaringstemperatur:Stue temperatur
- EINECS:212-682-9
Specifikationer
Specifikationstestresultater
- Solubility:800 g/l (25 °C)
- Molar Mass:291.29 g/mol
- Bulk Density:360 kg/m3
- pH:8.2 (10 g/l, H2O, 25 °C)
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



